1,1'-Hexamethylenebis[5-(2-ethylhex-1-yl)]biguanide dihydrochloride
Leverantör: Apollo Scientific
Varning
Synonyms: Alexidine dihydrochloride, N,N''''-Hexane-1,6-diylbis[N'-(2-ethylhex-1-yl)(imidodicarbonimidic diamide)] dihydrochloride
OR2503-1G OR2503-250MG
APOSOR2503-1GEA 2325 SEK
APOSOR2503-1G APOSOR2503-250MG
1,1'-Hexamethylenebis[5-(2-ethylhex-1-yl)]biguanide dihydrochloride
1,1'-Hexamethylenebis[5-(2-ethylhex-1-yl)]biguanide dihydrochloride
Has a antibacterial, antiplaque properties against Streptococcus mutans, Actinomyces ciscosus, and Actinomyces naesludii.
Formel: C₂₆H₅₆N₁₀·2ClH
MW: 581,712 g/mol
Smältpunkt: 220…224 °C
Storage Temperature: Frys
MDL Number: MFCD00082369
CAS nummer: 1715-30-6