4-[(1,1-Dioxidothiomorpholin-4-yl)methyl]benzoic acid
Leverantör: Apollo Scientific
Varning
Synonyms: 4-(4-Carboxybenzyl)thiomorpholine 1,1-dioxide
OR24489-1G OR24489-250MG
APOSOR24489-1GEA 570 SEK
APOSOR24489-1G APOSOR24489-250MG
4-[(1,1-Dioxidothiomorpholin-4-yl)methyl]benzoic acid
4-[(1,1-Dioxidothiomorpholin-4-yl)methyl]benzoic acid
Formel: C₁₂H₁₅NO₄S
MW: 269,317 g/mol
Smältpunkt: 222…223 °C
Storage Temperature: Omgivande
MDL Number: MFCD03086503
CAS nummer: 465514-21-0