4-(1,2,3-Thiadiazol-4-yl)benzyl bromide
Leverantör: Apollo Scientific
|
Varning
|
Synonyms:
4-[4-(Bromomethyl)phenyl]-1,2,3-thiadiazole
OR23262-1G
OR23262-250MG
APOSOR23262-1GEA
1525
SEK
APOSOR23262-1G
APOSOR23262-250MG
4-(1,2,3-Thiadiazol-4-yl)benzyl bromide
4-(1,2,3-Thiadiazol-4-yl)benzyl bromide
Formel:
C₉H₇BrN₂S MW: 255,134 g/mol Smältpunkt: 135…137 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00052102 CAS nummer: 163798-92-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...