(5-phenyl-1,3-oxazol-4-yl)methanol

Leverantör: Apollo Scientific

OR23140-1G OR23140-250MG
APOSOR23140-1GEA 3700 SEK
APOSOR23140-1G APOSOR23140-250MG
(5-phenyl-1,3-oxazol-4-yl)methanol
(5-phenyl-1,3-oxazol-4-yl)methanol
Formel: C₁₀H₉NO₂
MW: 175,184 g/mol
Smältpunkt: 94…95 °C
Storage Temperature: Omgivande
MDL Number: MFCD03086105
CAS nummer: 352018-88-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR