4-Nitrophenacylamine hydrochloride hydrate
Leverantör: Apollo Scientific
Synonyms: 2-Amino-1-(4-nitrophenyl)ethan-1-one hydrochloride hydrate
OR22661-1G OR22661-250MG
APOSOR22661-1GEA 2575 SEK
APOSOR22661-1G APOSOR22661-250MG
4-Nitrophenacylamine hydrochloride hydrate
4-Nitrophenacylamine hydrochloride hydrate
Formel: C₈H₈N₂O₃·ClH·H₂O
MW: 234.637 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD03659692
CAS nummer: 1049754-99-5