3-Hydroxy-1H-indazole 98%

Leverantör: Apollo Scientific
Varning

Synonyms: 1H-Indazol-3-ol

OR21771-1G OR21771-250MG
APOSOR21771-1GEA 2775 SEK
APOSOR21771-1G APOSOR21771-250MG
3-Hydroxy-1H-indazole 98%
3-Hydroxy-1H-indazole
Formel: C₇H₆N₂O
MW: 134,135 g/mol
Smältpunkt: 224…226 °C
Storage Temperature: Omgivande
MDL Number: MFCD00044658
CAS nummer: 100922-96-1
EINECS: 230-904-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR