Ethyl 2-iodo-5-methylbenzoate 95+%

Leverantör: Apollo Scientific

OR2175-10G OR2175-1G OR2175-5G
APOSOR2175-10GEA 1500 SEK
APOSOR2175-10G APOSOR2175-1G APOSOR2175-5G
Ethyl 2-iodo-5-methylbenzoate 95+%
Ethyl 2-iodo-5-methylbenzoate
Formel: C₁₀H₁₁IO₂
Storage Temperature: Omgivande
MDL Number: MFCD09998719
CAS nummer: 933585-44-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR