2-[(4-Chlorophenyl)thio]-3-nitropyridine
Leverantör: Apollo Scientific
OR21271-10G OR21271-1G OR21271-5G
APOSOR21271-10GEA 7450 SEK
APOSOR21271-10G APOSOR21271-1G APOSOR21271-5G
2-[(4-Chlorophenyl)thio]-3-nitropyridine
2-[(4-Chlorophenyl)thio]-3-nitropyridine
Formel: C₁₁H₇ClN₂O₂S
MW: 266,703 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00275701
CAS nummer: 26820-31-5