1-(4-Cyclohexylphenyl)-2-methylpropan-1-one 97+%
Leverantör: Apollo Scientific
Varning
OR2047-1G OR2047-5G
APOSOR2047-1GEA 1250 SEK
APOSOR2047-1G APOSOR2047-5G
1-(4-Cyclohexylphenyl)-2-methylpropan-1-one 97+%
1-(4-Cyclohexylphenyl)-2-methylpropan-1-one
Formel: C₁₆H₂₂O
Storage Temperature: Omgivande
MDL Number: MFCD01098369
CAS nummer: 53207-60-6