5-(Aminomethyl)-1,3-thiazole hydrochloride 95%
Leverantör: Apollo Scientific
Fara
Synonyms: Thiazol-5-ylmethanamine hydrochloride, (1,3-Thiazol-5-yl)methylamine hydrochloride
OR20040-1G OR20040-5G OR20040-10G
APOSOR20040-1GEA 1250 SEK
APOSOR20040-1G APOSOR20040-5G APOSOR20040-10G
5-(Aminomethyl)-1,3-thiazole hydrochloride 95%
5-(Aminomethyl)-1,3-thiazole hydrochloride
Formel: C₄H₆N₂S·ClH
MW: 150,63 g/mol
Smältpunkt: 173…181 °C
Storage Temperature: Kylskåp
MDL Number: MFCD06738808
CAS nummer: 131052-46-5