2-(Methylsulphanyl)benzamide
Leverantör: Apollo Scientific
OR200156-1G OR200156-250MG OR200156-500MG
APOSOR200156-1GEA 5675 SEK
APOSOR200156-1G APOSOR200156-250MG APOSOR200156-500MG
2-(Methylsulphanyl)benzamide
2-(Methylsulphanyl)benzamide
Formel: C₈H₉NOS
MW: 167,228 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00757485
CAS nummer: 54705-16-7