5-Ethenylpyrimidine
Leverantör: Apollo Scientific
|
Varning
|
OR18864-1G
OR18864-250MG
APOSOR18864-1GEA
7700
SEK
APOSOR18864-1G
APOSOR18864-250MG
5-Ethenylpyrimidine
5-Ethenylpyrimidine
Formel:
C₆H₆N₂ Kokpunkt: 86 °C (45 mm Hg) Storage Temperature: Kylskåp |
MDL Number:
MFCD18970848 CAS nummer: 53967-68-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...