5-Methylbenzo[b]thiophene-2-carboxylic acid

Leverantör: Apollo Scientific

OR183516-1G OR183516-250MG
APOSOR183516-1GEA 3500 SEK
APOSOR183516-1G APOSOR183516-250MG
5-Methylbenzo[b]thiophene-2-carboxylic acid
5-Methylbenzo[b]thiophene-2-carboxylic acid
Formel: C₁₀H₈O₂S
MW: 192,234 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD04969088
CAS nummer: 1505-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR