Methyl 2,6-dibromoisonicotinate
Leverantör: Apollo Scientific
Varning
Synonyms: Methyl 2,6-dibromopyridine-4-carboxylate, 2,6-Dibromo-4-(methoxycarbonyl)pyridine
OR18036-1G OR18036-5G
APOSOR18036-1GEA 975 SEK
APOSOR18036-1G APOSOR18036-5G
Methyl 2,6-dibromoisonicotinate
Methyl 2,6-dibromoisonicotinate
Formel: C₇H₅Br₂NO₂
MW: 294,928 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11042543
CAS nummer: 119308-57-5