2-Bromo-5,6-dihydrothieno[2,3-c]pyridin-7(4H)-one, tech
Leverantör: Apollo Scientific
Varning
Synonyms: 2-Bromo-7-oxo-4,5,6,7-tetrahydrothieno[2,3-c]pyridine
OR17555-1G OR17555-250MG
APOSOR17555-1GEA 10250 SEK
APOSOR17555-1G APOSOR17555-250MG
2-Bromo-5,6-dihydrothieno[2,3-c]pyridin-7(4H)-one, tech
2-Bromo-5,6-dihydrothieno[2,3-c]pyridin-7(4H)-one
Formel: C₇H₆BrNOS
MW: 232,098 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD14584992
CAS nummer: 960289-03-6