1-Benzyl-3-ethylpiperazine 95%

Leverantör: Apollo Scientific
Varning

OR16017-1G OR16017-250MG OR16017-5G
APOSOR16017-1GEA 1250 SEK
APOSOR16017-1G APOSOR16017-250MG APOSOR16017-5G
1-Benzyl-3-ethylpiperazine 95%
1-Benzyl-3-ethylpiperazine
Formel: C₁₃H₂₀N₂
Storage Temperature: Omgivande
MDL Number: MFCD03265492
CAS nummer: 313657-25-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR