1,3-Bis[(dimethylamino)methylene]thiourea

Leverantör: Apollo Scientific
Varning

Synonyms: 1,3-Bis[(dimethylamino)methylidene]thiourea

OR15284-1G OR15284-5G
APOSOR15284-1GEA 2025 SEK
APOSOR15284-1G APOSOR15284-5G
1,3-Bis[(dimethylamino)methylene]thiourea
1,3-Bis[(dimethylamino)methylene]thiourea
Formel: C₇H₁₄N₄S
MW: 186,278 g/mol
Smältpunkt: 187…189 °C
Storage Temperature: Omgivande
MDL Number: MFCD00173502
CAS nummer: 96818-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR