1-[(Thien-2-yl)methyl]piperidine-4-carboxaldehyde 97%
Leverantör: Apollo Scientific
Fara
Synonyms: 4-Formyl-1-[(thien-2-yl)methyl]piperidine, 2-[(4-Formylpiperidin-1-yl)methyl]thiophene
OR14936-250MG OR14936-1G
APOSOR14936-250MGEA 1400 SEK
APOSOR14936-250MG APOSOR14936-1G
1-[(Thien-2-yl)methyl]piperidine-4-carboxaldehyde 97%
1-[(Thien-2-yl)methyl]piperidine-4-carboxaldehyde
Formel: C₁₁H₁₅NOS
MW: 209,308 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09879953
CAS nummer: 926921-80-4
UN: 2735
ADR: 8,III