1-Methyl-4-nitro-1H-pyrazole-5-carboxylic acid
Leverantör: Apollo Scientific
OR13452-1G OR13452-25G OR13452-5G
APOSOR13452-1GEA 1700 SEK
APOSOR13452-1G APOSOR13452-25G APOSOR13452-5G
1-Methyl-4-nitro-1H-pyrazole-5-carboxylic acid
1-Methyl-4-nitro-1H-pyrazole-5-carboxylic acid
Formel: C₅H₅N₃O₄
MW: 171,111 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00222738
CAS nummer: 92534-69-5