2-Phenyl-1-(piperazin-1-yl)ethan-1-one
Leverantör: Apollo Scientific
Varning
Synonyms: [2-Oxo-2-(piperazin-1-yl)ethyl]benzene, 1-(Phenylacetyl)piperazine
OR13308-100MG OR13308-1G OR13308-250MG
APOSOR13308-100MGEA 2925 SEK
APOSOR13308-100MG APOSOR13308-1G APOSOR13308-250MG
2-Phenyl-1-(piperazin-1-yl)ethan-1-one
2-Phenyl-1-(piperazin-1-yl)ethan-1-one
Formel: C₁₂H₁₆N₂O
MW: 204,268 g/mol
Kokpunkt: 190…191 °C (17 mm Hg)
Storage Temperature: Kylskåp
MDL Number: MFCD01566912
CAS nummer: 88372-33-2