2-[(Pyrazin-2-yl)oxy]benzaldehyde
Leverantör: Apollo Scientific
Varning
Synonyms: 2-(Pyrazin-2-yloxy)benzaldehyde
OR12896-100MG OR12896-250MG OR12896-1G
APOSOR12896-100MGEA 1700 SEK
APOSOR12896-100MG APOSOR12896-250MG APOSOR12896-1G
2-[(Pyrazin-2-yl)oxy]benzaldehyde
2-[(Pyrazin-2-yl)oxy]benzaldehyde
Formel: C₁₁H₈N₂O₂
MW: 200.193 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD03840028
CAS nummer: 887344-43-6