3-(2-Chlorophenyl)morpholine hydrochloride
Leverantör: Apollo Scientific
Varning
Synonyms: 1-Chloro-2-(morpholin-3-yl)benzene hydrochloride
OR12857-100MG OR12857-250MG
APOSOR12857-100MGEA 4475 SEK
APOSOR12857-100MG APOSOR12857-250MG
3-(2-Chlorophenyl)morpholine hydrochloride
3-(2-Chlorophenyl)morpholine hydrochloride
Formel: C₁₀H₁₂ClNO·ClH
MW: 234,122 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD03840094
CAS nummer: 1172507-31-1