4-Iodo-4'-methyldiphenyl ether
Leverantör: Apollo Scientific
Varning
Synonyms: 4-(4-Iodophenoxy)toluene, 1-Iodo-4-(4-methylphenoxy)benzene
OR11364-1G OR11364-250MG
APOSOR11364-1GEA 2025 SEK
APOSOR11364-1G APOSOR11364-250MG
4-Iodo-4'-methyldiphenyl ether
4-Iodo-4'-methyldiphenyl ether
Formel: C₁₃H₁₁IO
MW: 310,13 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD06796301
CAS nummer: 1017793-91-7