4-Hydroxy-7-methoxyindane
Leverantör: Apollo Scientific
Varning
OR11192-1G OR11192-250MG
APOSOR11192-1GEA 4525 SEK
APOSOR11192-1G APOSOR11192-250MG
4-Hydroxy-7-methoxyindane
4-Hydroxy-7-methoxyindane
Formel: C₁₀H₁₂O₂
Storage Temperature: Omgivande
MDL Number: MFCD04038952
CAS nummer: 38998-04-8