4-Oxo-6-phenyl-2-thiOxo-1,2,3,4-tetrahydropyrimidine-5-carbonitrile

Leverantör: Apollo Scientific

OR111408-10G OR111408-1G OR111408-5G
APOSOR111408-10GEA 0 SEK
APOSOR111408-10G APOSOR111408-1G APOSOR111408-5G
4-Oxo-6-phenyl-2-thiOxo-1,2,3,4-tetrahydropyrimidine-5-carbonitrile
4-Oxo-6-phenyl-2-thiOxo-1,2,3,4-tetrahydropyrimidine-5-carbonitrile
Formel: C₁₁H₇N₃OS
MW: 229,258 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00228595
CAS nummer: 70638-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR