4-(1,1-Dimethoxyethyl)pyrimidin-2-amine
Leverantör: Apollo Scientific
OR111311-1G OR111311-250MG OR111311-500MG
APOSOR111311-1GEA 10300 SEK
APOSOR111311-1G APOSOR111311-250MG APOSOR111311-500MG
4-(1,1-Dimethoxyethyl)pyrimidin-2-amine
4-(1,1-Dimethoxyethyl)pyrimidin-2-amine
Formel: C₈H₁₃N₃O₂
Storage Temperature: Omgivande
MDL Number: MFCD16988356
CAS nummer: 106157-85-1