N'-(2,4-Dichloro-6-methylphenyl)-N,N-diisopropylthiourea
Leverantör: Apollo Scientific
OR111140-10G OR111140-1G OR111140-5G
APOSOR111140-10GEA 0 SEK
APOSOR111140-10G APOSOR111140-1G APOSOR111140-5G
N'-(2,4-Dichloro-6-methylphenyl)-N,N-diisopropylthiourea
N'-(2,4-Dichloro-6-methylphenyl)-N,N-diisopropylthiourea
Formel: C₁₄H₂₀Cl₂N₂S
MW: 319,293 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD28987632
CAS nummer: 1980044-24-3