2-Cyano-n-(3-methoxybenzyl)acetamide

Leverantör: Apollo Scientific
Varning

Synonyms: 2-Cyano-N-(3-methoxybenzyl)acetamide

OR111057-1G OR111057-5G
APOSOR111057-1GEA 3450 SEK
APOSOR111057-1G APOSOR111057-5G
2-Cyano-n-(3-methoxybenzyl)acetamide
2-Cyano-n-(3-methoxybenzyl)acetamide
Formel: C₁₁H₁₂N₂O₂
MW: 204,225 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD12824545
CAS nummer: 566926-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR