2-(3,5-Dichlorophenyl)hydrazinecarbothioamide, Technical Grade
Leverantör: Apollo Scientific
Varning
OR111045-10G OR111045-1G OR111045-5G
APOSOR111045-10GEA 500 SEK
APOSOR111045-10G APOSOR111045-1G APOSOR111045-5G
2-(3,5-Dichlorophenyl)hydrazinecarbothioamide, Technical Grade
2-(3,5-Dichlorophenyl)hydrazinecarbothioamide
Formel: C₇H₇Cl₂N₃S
MW: 236,122 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD03305525
CAS nummer: 96423-39-1