N-(2-Bromo-4-isopropylphenyl)thiourea
Leverantör: Apollo Scientific
OR111042-1G OR111042-5G
APOSOR111042-1GEA 0 SEK
APOSOR111042-1G APOSOR111042-5G
N-(2-Bromo-4-isopropylphenyl)thiourea
N-(2-Bromo-4-isopropylphenyl)thiourea
Formel: C₁₀H₁₃BrN₂S
MW: 273.193 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD18379866
CAS nummer: 724426-94-2