1-Bromo-2-(2,2-diethoxyethoxy)benzene
Leverantör: Apollo Scientific
OR110847-1G OR110847-5G OR110847-10G
APOSOR110847-1GEA 2200 SEK
APOSOR110847-1G APOSOR110847-5G APOSOR110847-10G
1-Bromo-2-(2,2-diethoxyethoxy)benzene
1-Bromo-2-(2,2-diethoxyethoxy)benzene
Formel: C₁₂H₁₇BrO₃
MW: 289.166 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD18848576
CAS nummer: 253429-15-1