4,6-Dimethyl-2-thien-2-ylnicotinonitrile
Leverantör: Apollo Scientific
OR110670-1G OR110670-5G
APOSOR110670-1GEA 2475 SEK
APOSOR110670-1G APOSOR110670-5G
4,6-Dimethyl-2-thien-2-ylnicotinonitrile
4,6-Dimethyl-2-thien-2-ylnicotinonitrile
Formel: C₁₂H₁₀N₂S
MW: 214,286 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD21602387
CAS nummer: 1427460-28-3