2-(3,4-Dimethylphenoxy)benzaldehyde

Leverantör: Apollo Scientific

OR110498-1G OR110498-5G
APOSOR110498-1GEA 2075 SEK
APOSOR110498-1G APOSOR110498-5G
2-(3,4-Dimethylphenoxy)benzaldehyde
2-(3,4-Dimethylphenoxy)benzaldehyde
Formel: C₁₅H₁₄O₂
MW: 226,27 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11115770
CAS nummer: 289718-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR