1-(1H-Indol-3-ylmethyl)piperidine-4-carboxylic acid
Leverantör: Apollo Scientific
Varning
OR110381-1G OR110381-250MG
APOSOR110381-1GEA 3325 SEK
APOSOR110381-1G APOSOR110381-250MG
1-(1H-Indol-3-ylmethyl)piperidine-4-carboxylic acid
1-(1H-Indol-3-ylmethyl)piperidine-4-carboxylic acid
Formel: C₁₅H₁₈N₂O₂
MW: 258,316 g/mol
Smältpunkt: 228 °C (dec.)
Storage Temperature: Kylskåp
MDL Number: MFCD06739714
CAS nummer: 100957-76-4