Ethyl 4-methyl-3-nitrobenzoate
Leverantör: Apollo Scientific
Varning
OR110364-1G OR110364-5G
APOSOR110364-1GEA 480 SEK
APOSOR110364-1G APOSOR110364-5G
Ethyl 4-methyl-3-nitrobenzoate
Ethyl 4-methyl-3-nitrobenzoate
Formel: C₁₀H₁₁NO₄
Storage Temperature: Omgivande
MDL Number: MFCD06204349
CAS nummer: 19013-15-1