Ethyl 2-amino-3-methylbenzoate

Leverantör: Apollo Scientific

OR110353-10G OR110353-1G OR110353-5G
APOSOR110353-10GEA 5825 SEK
APOSOR110353-10G APOSOR110353-1G APOSOR110353-5G
Ethyl 2-amino-3-methylbenzoate
Ethyl 2-amino-3-methylbenzoate
Formel: C₁₀H₁₃NO₂
MW: 179,216 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD11183528
CAS nummer: 64818-79-7
UN: 3259
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR