6-Hydrazinonicotinonitrile 95%

Leverantör: Apollo Scientific
Varning

OR110312-1G OR110312-250MG
APOSOR110312-1GEA 1500 SEK
APOSOR110312-1G APOSOR110312-250MG
6-Hydrazinonicotinonitrile 95%
6-Hydrazinonicotinonitrile
Formel: C₆H₆N₄
Smältpunkt: 181…183 °C
Storage Temperature: Omgivande
MDL Number: MFCD09040676
CAS nummer: 104408-24-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR