2-(5-Amino-3-phenyl-1H-pyrazol-1-yl)ethanol
Leverantör: Apollo Scientific
OR110311-1G OR110311-250MG OR110311-5G
APOSOR110311-1GEA 2950 SEK
APOSOR110311-1G APOSOR110311-250MG APOSOR110311-5G
2-(5-Amino-3-phenyl-1H-pyrazol-1-yl)ethanol
2-(5-Amino-3-phenyl-1H-pyrazol-1-yl)ethanol
Formel: C₁₁H₁₃N₃O
MW: 203.24 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD05263730
CAS nummer: 14085-42-8