3-(Dimethylamino)-1-(3-nitrophenyl)prop-2-en-1-one
Leverantör: Apollo Scientific
OR110306-1G OR110306-5G
APOSOR110306-1GEA 1025 SEK
APOSOR110306-1G APOSOR110306-5G
3-(Dimethylamino)-1-(3-nitrophenyl)prop-2-en-1-one
3-(Dimethylamino)-1-(3-nitrophenyl)prop-2-en-1-one
Formel: C₁₁H₁₂N₂O₃
MW: 220.225 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00024513
CAS nummer: 115955-48-1