Ethyl 4-octylbenzoate

Leverantör: Apollo Scientific

OR110268-1G OR110268-5G
APOSOR110268-1GEA 1525 SEK
APOSOR110268-1G APOSOR110268-5G
Ethyl 4-octylbenzoate
Ethyl 4-octylbenzoate
Formel: C₁₇H₂₆O₂
Storage Temperature: Omgivande
MDL Number: MFCD06204568
CAS nummer: 133002-71-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR