1,3-Benzoxazole-7-carboxylic acid
Leverantör: Apollo Scientific
|
Varning
|
Synonyms:
7-Carboxy-1,3-benzoxazole, Benz[d]oxazole-7-carboxylic acid
OR110149-1G
OR110149-250MG
APOSOR110149-1GEA
5600
SEK
APOSOR110149-1G
APOSOR110149-250MG
1,3-Benzoxazole-7-carboxylic acid
1,3-Benzoxazole-7-carboxylic acid
Formel:
C₈H₅NO₃ MW: 163,13 g/mol Smältpunkt: 240…250 °C (subl.) Storage Temperature: Omgivande |
MDL Number:
MFCD11890785 CAS nummer: 208772-24-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...