5-Chloro-2-(1-methylhydrazino)pyridine

Leverantör: Apollo Scientific
Varning

OR110063-1G OR110063-10G OR110063-5G
APOSOR110063-1GEA 175 SEK
APOSOR110063-1G APOSOR110063-10G APOSOR110063-5G
5-Chloro-2-(1-methylhydrazino)pyridine
5-Chloro-2-(1-methylhydrazino)pyridine
Formel: C₆H₈ClN₃
MW: 157,601 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09999898
CAS nummer: 1378830-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR