Methyl 1-methyl-1H-indole-2-carboxylate
Leverantör: Apollo Scientific
Varning
OR110042-1G OR110042-5G
APOSOR110042-1GEA 720 SEK
APOSOR110042-1G APOSOR110042-5G
Methyl 1-methyl-1H-indole-2-carboxylate
Methyl 1-methyl-1H-indole-2-carboxylate
Formel: C₁₁H₁₁NO₂
Storage Temperature: Omgivande
MDL Number: MFCD00455976
CAS nummer: 37493-34-8