[3-(Pyridin-2-yloxy)phenyl]methanol
Leverantör: Apollo Scientific
|
Varning
|
OR110006-1G
OR110006-250MG
OR110006-5G
APOSOR110006-1GEA
2150
SEK
APOSOR110006-1G
APOSOR110006-250MG
APOSOR110006-5G
[3-(Pyridin-2-yloxy)phenyl]methanol
[3-(Pyridin-2-yloxy)phenyl]methanol
Formel:
C₁₂H₁₁NO₂ Smältpunkt: 56…59 °C Storage Temperature: Omgivande |
MDL Number:
MFCD09025855 CAS nummer: 869901-22-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...