Sodium 3-chlorobenzoate
Leverantör: Apollo Scientific
Varning
OR10911X-25G OR10911X-50G OR10911X-5G
APOSOR10911X-25GEA 2325 SEK
APOSOR10911X-25G APOSOR10911X-50G APOSOR10911X-5G
Sodium 3-chlorobenzoate
Sodium 3-chlorobenzoate
Formel: C₇H₄ClO₂·Na
MW: 178,548 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD08059569
CAS nummer: 17264-88-9