Ethyl [1,1'-biphenyl]-3-carboxylate
Leverantör: Apollo Scientific
Varning
OR10603-1G OR10603-250MG
APOSOR10603-1GEA 550 SEK
APOSOR10603-1G APOSOR10603-250MG
Ethyl [1,1'-biphenyl]-3-carboxylate
Ethyl [1,1'-biphenyl]-3-carboxylate
Formel: C₁₅H₁₄O₂
Storage Temperature: Omgivande
MDL Number: MFCD00671950
CAS nummer: 19926-50-2