2,3-Dihydrobenzo[b]thiophene-6-sulphonyl chloride 1,1-dioxide
Leverantör: Apollo Scientific
Fara
OR103628-1G OR103628-250MG OR103628-5G
APOSOR103628-1GEA 3225 SEK
APOSOR103628-1G APOSOR103628-250MG APOSOR103628-5G
2,3-Dihydrobenzo[b]thiophene-6-sulphonyl chloride 1,1-dioxide
2,3-Dihydrobenzo[b]thiophene-6-sulphonyl chloride 1,1-dioxide
Formel: C₈H₇ClO₄S₂
MW: 266,722 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD06808708
CAS nummer: 208643-45-2