2-(1H-Imidazol-1-yl)benzoic acid
Leverantör: Apollo Scientific
Varning
Synonyms: 1-(2-Carboxyphenyl)-1H-imidazole, 2-(1-Imidazolyl)benzoic Acid
OR0715-1G OR0715-250MG OR0715-5G
APOSOR0715-1GEA 1050 SEK
APOSOR0715-1G APOSOR0715-250MG APOSOR0715-5G
2-(1H-Imidazol-1-yl)benzoic acid
2-(1H-Imidazol-1-yl)benzoic acid
Formel: C₁₀H₈N₂O₂
MW: 188.183 g/mol
Smältpunkt: 198…200 °C
Storage Temperature: Omgivande
MDL Number: MFCD06254507
CAS nummer: 159589-67-0