Ethyl 3-(chlorosulphonyl)isonicotinate 95%
Leverantör: Apollo Scientific
Fara
Synonyms: Ethyl 3-(chlorosulphonyl)pyridine-4-carboxylate
OR0448-1G OR0448-250MG OR0448-50MG
APOSOR0448-1GEA 3875 SEK
APOSOR0448-1G APOSOR0448-250MG APOSOR0448-50MG
Ethyl 3-(chlorosulphonyl)isonicotinate 95%
Ethyl 3-(chlorosulphonyl)isonicotinate
Formel: C₈H₈ClNO₄S
MW: 249,671 g/mol
Kokpunkt: 72…78 °C (0,1 mm Hg)
Storage Temperature: Omgivande
MDL Number: MFCD01313855
CAS nummer: 306936-12-9
UN: 3265
ADR: 8,III