3-(tert-Butyl)phenylhydrazine hydrochloride, Technical Grade
Leverantör: Apollo Scientific
Varning
Synonyms: (3-(tert-Butyl)phenyl)hydrazine hydrochloride, 1-(tert-Butyl)-3-hydrazinobenzene
OR016187-1G OR016187-250MG OR016187-5G
APOSOR016187-1GEA 2575 SEK
APOSOR016187-1G APOSOR016187-250MG APOSOR016187-5G
3-(tert-Butyl)phenylhydrazine hydrochloride, Technical Grade
3-(tert-Butyl)phenylhydrazine hydrochloride
Formel: C₁₀H₁₆N₂·ClH
MW: 200.708 g/mol
Smältpunkt: 146…148 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00833404
CAS nummer: 306937-27-9
UN: 2811
ADR: 6.1 III